Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetraethylammonium Hydroxide Solution

Catalog No. 177806100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-780-6100G 100 g
17-780-6500G 500 g
17-780-65KG 5 kg
17-780-61KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 17-780-6100G Supplier Sigma Aldrich Supplier No. 177806100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The product is 20wt.% solution of tetraethylammonium hydroxide (TEA hydroxide, TENOH) in H2O. TEA hydroxide is a quaternary ammonium hydroxide (QAH). It participates in the synthesis of zeolite beta. TENOH peptized titania aqueous sols have been employed in the preparation of nanocrystalline anatase films.

Specifications

CAS 77-98-5
Density 1.01 g/mL at 25°C
Molecular Formula C8H21NO
Refractive Index n20/D 1.373
Linear Formula (C2H5)4N(OH)
MDL Number MFCD00009024
Quantity 100 g
Synonym TEA hydroxide
Molecular Weight (g/mol) 147.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.