Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetraethylammonium nitrate | 1941-26-0 | MFCD00041978 | ≥98.0% | 192.26 g/mol | C8H20N2O3 | 10 G
SDP

Catalog No. 5000283644
Encompass_Preferred

Tetraethylammonium nitrate is a quaternary ammonium salt supplied as a high-purity solid reagent for research use. It is used as an electrolyte and reagent in biochemical assays and electrochemical applications; molecular weight 192.26 g/mol. The material is typically a white crystalline powder with reported purity ≥98% and is soluble in DMSO (~10 mM).

  • High-purity reagent suitable for laboratory assays.
  • Functions as a quaternary ammonium electrolyte in electrochemistry.
  • Soluble in DMSO for preparation of stock solutions.
  • White crystalline powder for easy weighing and handling.
  • Molecular weight provided for stoichiometric calculations.

Catalog No. 50-002-83644 Supplier Medchemexpress LLC Supplier No. HYW12767710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetraethylammonium nitrate is a quaternary ammonium salt supplied as a high-purity solid reagent for research use. It is used as an electrolyte and reagent in biochemical assays and electrochemical applications; molecular weight 192.26 g/mol. The material is typically a white crystalline powder with reported purity ≥98% and is soluble in DMSO (~10 mM).

  • High-purity reagent suitable for laboratory assays.
  • Functions as a quaternary ammonium electrolyte in electrochemistry.
  • Soluble in DMSO for preparation of stock solutions.
  • White crystalline powder for easy weighing and handling.
  • Molecular weight provided for stoichiometric calculations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.