Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetraethylammonium nitrate | 1941-26-0 | MFCD00041978 | 99.9% | 192.26 g/mol | C8H20N2O3 | 1 G
SDP

Catalog No. 5000283607
Encompass_Preferred

Tetraethylammonium nitrate is a quaternary ammonium salt supplied as a high-purity solid for laboratory research and biochemical assays. It is commonly used as an electrolyte and reagent in chemical and electrochemical applications and is supplied in 1 g packaging.

  • High purity (~99.9%) suitable for analytical and research use.
  • Quaternary ammonium nitrate salt, solid form.
  • CAS number 1941-26-0 for unambiguous identification.
  • Molecular weight 192.26 g/mol.
  • Useful as an electrolyte and reagent in electrochemistry and synthesis.
  • Convenient 1 g packaging for small-scale experiments.

Catalog No. 50-002-83607 Supplier Medchemexpress LLC Supplier No. HYW1276771G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetraethylammonium nitrate is a quaternary ammonium salt supplied as a high-purity solid for laboratory research and biochemical assays. It is commonly used as an electrolyte and reagent in chemical and electrochemical applications and is supplied in 1 g packaging.

  • High purity (~99.9%) suitable for analytical and research use.
  • Quaternary ammonium nitrate salt, solid form.
  • CAS number 1941-26-0 for unambiguous identification.
  • Molecular weight 192.26 g/mol.
  • Useful as an electrolyte and reagent in electrochemistry and synthesis.
  • Convenient 1 g packaging for small-scale experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.