Learn More
Medchemexpress LLC Tetraethylammonium nitrate | 1941-26-0 | MFCD00041978 | 99.9% | 192.26 g/mol | C8H20N2O3 | 100 G
SDP
Catalog No. 5000282782
Encompass_Preferred
Tetraethylammonium nitrate is a quaternary ammonium salt supplied as a white crystalline powder for research use. It is commonly used as a supporting electrolyte and reagent in electrochemical and synthetic applications. Key properties include CAS 1941-26-0, molecular weight 192.26 g/mol, and high assay purity suitable for laboratory work.
  • High purity (~99.9%) suitable for analytical and research applications.
  • Serves as a supporting electrolyte in electrochemistry.
  • White crystalline powder, convenient to weigh and handle.
  • Soluble in polar solvents; consult solubility data for specifics.
  • Stable when stored under recommended conditions for long-term use.
  • Available in laboratory pack sizes up to 100 G.
Catalog No. 50-002-82782 Supplier Medchemexpress LLC Supplier No. HYW127677100G
Tetraethylammonium nitrate is a quaternary ammonium salt supplied as a white crystalline powder for research use. It is commonly used as a supporting electrolyte and reagent in electrochemical and synthetic applications. Key properties include CAS 1941-26-0, molecular weight 192.26 g/mol, and high assay purity suitable for laboratory work.
  • High purity (~99.9%) suitable for analytical and research applications.
  • Serves as a supporting electrolyte in electrochemistry.
  • White crystalline powder, convenient to weigh and handle.
  • Soluble in polar solvents; consult solubility data for specifics.
  • Stable when stored under recommended conditions for long-term use.
  • Available in laboratory pack sizes up to 100 G.