Learn More
Medchemexpress LLC Tetraethylammonium nitrate | 1941-26-0 | MFCD00041978 | 99.9% | 192.26 g/mol | C8H20N2O3 | 5 G
SDP
Catalog No. 5000283516
Encompass_Preferred
Tetraethylammonium nitrate is a quaternary ammonium salt supplied as a solid reagent for laboratory research. It is commonly used as an electrolyte and biochemical assay reagent and is provided with quality documentation for safe handling and traceable results.
  • High purity 99.87% for reliable experimental results.
  • Solid powder form for easy handling and storage.
  • Useful as an electrolyte and biochemical assay reagent.
  • Documented with COA and SDS for traceability and safety.
  • Stable under recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years).
Catalog No. 50-002-83516 Supplier Medchemexpress LLC Supplier No. HYW1276775G
Tetraethylammonium nitrate is a quaternary ammonium salt supplied as a solid reagent for laboratory research. It is commonly used as an electrolyte and biochemical assay reagent and is provided with quality documentation for safe handling and traceable results.
  • High purity 99.87% for reliable experimental results.
  • Solid powder form for easy handling and storage.
  • Useful as an electrolyte and biochemical assay reagent.
  • Documented with COA and SDS for traceability and safety.
  • Stable under recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years).