Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetraethylammonium perchlorate | 2567-83-1 | 98.0% | 229.70 | 25 G
SDP

Catalog No. 5000451780
Encompass_Preferred

Tetraethylammonium perchlorate is an organic compound commonly used as an electrolyte in electrochemistry and as a source of perchlorate anion in analytical chemistry. It has various applications in the production of batteries, fuel cells, and electronic components due to its high ionic conductivity and stability under extreme conditions. It can also be used as a reference standard in analytical methods such as ion chromatography.

  • Used as an electrolyte in electrochemistry
  • Source of perchlorate anion in analytical chemistry
  • High ionic conductivity and stability under extreme conditions
  • Applications in batteries, fuel cells, and electronic components
  • Can be used as a reference standard in analytical methods

Catalog No. 50-004-51780 Supplier Medchemexpress LLC Supplier No. 25GHYW009235
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetraethylammonium perchlorate is an organic compound commonly used as an electrolyte in electrochemistry and as a source of perchlorate anion in analytical chemistry. It has various applications in the production of batteries, fuel cells, and electronic components due to its high ionic conductivity and stability under extreme conditions. It can also be used as a reference standard in analytical methods such as ion chromatography.

  • Used as an electrolyte in electrochemistry
  • Source of perchlorate anion in analytical chemistry
  • High ionic conductivity and stability under extreme conditions
  • Applications in batteries, fuel cells, and electronic components
  • Can be used as a reference standard in analytical methods

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.