Learn More
Medchemexpress LLC Tetraethylammonium perchlorate | 2567-83-1 | MFCD00041977 | ≥98.0% | 229.70 g/mol | C8H20ClNO4 | 1 G
SDP
Catalog No. 5000282665
Encompass_Preferred
Tetraethylammonium perchlorate is a quaternary ammonium salt used as an electrolyte and analytical reagent. It provides the tetraethylammonium cation paired with the perchlorate anion, offering high ionic conductivity and good stability for research and analytical workflows. Common applications include electrochemical studies, battery and fuel cell research, and use as reference standards in chromatography.
  • High purity (≥98.0%) suitable for analytical and research use.
  • Provides a stable, highly conductive electrolyte for electrochemical studies.
  • Compatible with preparation of stock solutions for ion chromatography and other analyses.
  • Available in small pack sizes convenient for laboratory experiments.
  • Molecular weight 229.70 g/mol and formula C8H20ClNO4 for accurate stoichiometry.
Catalog No. 50-002-82665 Supplier Medchemexpress LLC Supplier No. HYW1276761G
Tetraethylammonium perchlorate is a quaternary ammonium salt used as an electrolyte and analytical reagent. It provides the tetraethylammonium cation paired with the perchlorate anion, offering high ionic conductivity and good stability for research and analytical workflows. Common applications include electrochemical studies, battery and fuel cell research, and use as reference standards in chromatography.
  • High purity (≥98.0%) suitable for analytical and research use.
  • Provides a stable, highly conductive electrolyte for electrochemical studies.
  • Compatible with preparation of stock solutions for ion chromatography and other analyses.
  • Available in small pack sizes convenient for laboratory experiments.
  • Molecular weight 229.70 g/mol and formula C8H20ClNO4 for accurate stoichiometry.