Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylammonium Perchlorate 95%, Thermo Scientific™

Catalog No. AC420331000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC420331000 100 g
AC420330250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC420331000 Supplier Thermo Scientific Chemicals Supplier No. 420331000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2567-83-1
Molecular Formula C8H20ClNO4
Molecular Weight (g/mol) 229.70
MDL Number MFCD00041977
InChI Key WGHUNMFFLAMBJD-UHFFFAOYSA-M
Synonym tetraethylammonium perchlorate, ethanaminium, n,n,n-triethyl-, perchlorate, tetraethylazanium perchlorate, ethanaminium, n,n,n-triethyl-, perchlorate 1:1, ammonium, tetraethyl-, perchlorate, tetraethylammoniumperchlorate, acmc-1ccq7, ksc203k4p, tetraethyl ammonium perchlorate, tetraethylammoniumperchlorate, dry
PubChem CID 2734932
IUPAC Name tetraethylazanium perchlorate
SMILES [O-][Cl](=O)(=O)=O.CC[N+](CC)(CC)CC

Specifications

Color White
Infrared Spectrum Authentic
Linear Formula (CH5)4NClO4
Quantity 100 g
Formula Weight 229.7
Percent Purity ≥94%
Physical Form Crystalline Powder
Chemical Name or Material Tetraethylammonium perchlorate, 95%
EINECSNumber 219-904-3
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.