Learn More
Medchemexpress LLC Tetraethylammonium tetrafluoroborate | 429-06-1 | MFCD00011827 | >98.0% | 217.06 g/mol | C8H20BF4N | 10 G
SDP
Catalog No. 5000302093
Encompass_Preferred
Tetraethylammonium tetrafluoroborate is a quaternary ammonium salt used as a reagent and electrolyte in chemical and biochemical research. It provides tetraethylammonium cations and tetrafluoroborate anions for phase-transfer catalysis, organic synthesis, electrochemical studies, and pharmacological/biochemical experiments.
  • High purity ≥98.0% supports reproducible experimental results.
  • White solid with high thermal stability for reliable handling and storage.
  • Soluble in common organic solvents and effective as a supporting electrolyte.
  • Functions as a phase-transfer catalyst and source of tetraethylammonium ions.
  • Available in small pack sizes suitable for laboratory-scale use.
Catalog No. 50-003-02093 Supplier Medchemexpress LLC Supplier No. HYW01410610G
Tetraethylammonium tetrafluoroborate is a quaternary ammonium salt used as a reagent and electrolyte in chemical and biochemical research. It provides tetraethylammonium cations and tetrafluoroborate anions for phase-transfer catalysis, organic synthesis, electrochemical studies, and pharmacological/biochemical experiments.
  • High purity ≥98.0% supports reproducible experimental results.
  • White solid with high thermal stability for reliable handling and storage.
  • Soluble in common organic solvents and effective as a supporting electrolyte.
  • Functions as a phase-transfer catalyst and source of tetraethylammonium ions.
  • Available in small pack sizes suitable for laboratory-scale use.