Learn More
Medchemexpress LLC Tetraethylammonium tetrafluoroborate | 429-06-1 | MFCD00011827 | ≥98.0% | 217.06 g/mol | C8H20BF4N | 100 G
SDP
Supplier:  Medchemexpress LLC HYW014106100G
Encompass_Preferred
Tetraethylammonium tetrafluoroborate is a quaternary ammonium salt (CAS 429-06-1) supplied as a solid reagent for research and synthesis. It is commonly used as a phase-transfer catalyst, an electrolyte in electrochemical applications, and as a source of tetraethylammonium ions in organic and biochemical experiments. Typical purity is ≥98.0% and molecular weight is 217.06 g/mol.
  • High purity suitable for research applications.
  • Useful as a phase-transfer catalyst in organic synthesis.
  • Functions as an electrolyte in electrochemical cells and studies.
  • Solid form with convenient laboratory handling and storage.
  • Provides a reliable source of tetraethylammonium ions for biochemical experiments.
Catalog No. 50-003-04095