Learn More
Medchemexpress LLC Tetraethylammonium tetrafluoroborate | 429-06-1 | MFCD00011827 | 98.0% | 217.06 g/mol | C8H20BF4N | 25 G
SDP
Catalog No. 5000303505
Encompass_Preferred
Tetraethylammonium tetrafluoroborate is a quaternary ammonium salt used as a supporting electrolyte in non-aqueous electrochemistry and as a phase-transfer catalyst in organic synthesis. It is supplied as a solid reagent with high purity for laboratory research and synthetic applications.
  • High purity (98.0%) suitable for analytical and synthetic work.
  • Solid form, easy to weigh and handle in laboratory settings.
  • Compatible as a supporting electrolyte in non-aqueous electrochemistry.
  • Effective phase-transfer catalyst for a range of organic transformations.
  • Available in multiple pack sizes for small-scale and bulk use.
Catalog No. 50-003-03505 Supplier Medchemexpress LLC Supplier No. 25GHYW014106
Tetraethylammonium tetrafluoroborate is a quaternary ammonium salt used as a supporting electrolyte in non-aqueous electrochemistry and as a phase-transfer catalyst in organic synthesis. It is supplied as a solid reagent with high purity for laboratory research and synthetic applications.
  • High purity (98.0%) suitable for analytical and synthetic work.
  • Solid form, easy to weigh and handle in laboratory settings.
  • Compatible as a supporting electrolyte in non-aqueous electrochemistry.
  • Effective phase-transfer catalyst for a range of organic transformations.
  • Available in multiple pack sizes for small-scale and bulk use.