Learn More
Medchemexpress LLC Tetraethylammonium tetrafluoroborate | 429-06-1 | MFCD00011827 | ≥98.0% | 217.06 g·mol⁻¹ | C8H20BF4N | 50 G
SDP
Catalog No. 5000303856
Encompass_Preferred
Tetraethylammonium tetrafluoroborate is a quaternary ammonium salt used in organic synthesis and electrochemical research. It is employed where a non-nucleophilic, bulky tetraalkylammonium counterion is required, serving as a reagent, phase-transfer catalyst, and supporting electrolyte.
Catalog No. 50-003-03856 Supplier Medchemexpress LLC Supplier No. HYW01410650G
Tetraethylammonium tetrafluoroborate is a quaternary ammonium salt used in organic synthesis and electrochemical research. It is employed where a non-nucleophilic, bulky tetraalkylammonium counterion is required, serving as a reagent, phase-transfer catalyst, and supporting electrolyte.