Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetraethylammonium tetrafluoroborate | 429-06-1 | MFCD00011827 | ≥99.0% | 217.06 g/mol | C8H20BF4N | 5 G
SDP

Supplier:  Medchemexpress LLC HYW0141065G

Encompass_Preferred

Tetraethylammonium tetrafluoroborate is the tetrafluoroborate salt of the tetraethylammonium cation, supplied as a white, hygroscopic crystalline powder. It is used as a supporting electrolyte, phase-transfer reagent, and source of tetraalkylammonium cations in organic and electrochemical synthesis. Handle with appropriate protective measures because of irritant and aquatic toxicity hazards.

  • CAS number 429-06-1.
  • Molecular weight 217.06 g/mol.
  • Chemical formula C8H20BF4N.
  • Physical form white, hygroscopic crystalline powder.
  • Typical uses include supporting electrolyte, phase-transfer reagent, and reagent in electrochemical and organic synthesis.
  • Purity ≥99.0% (assay by titration).
  • Hazards include skin and eye irritation and toxicity to aquatic life.

Catalog No. 50-003-04122


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.