Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetraethylammonium trifluoromethanesulfonate | 35895-69-3 | MFCD00042586 | 98.0% | 279.32 g·mol⁻¹ | C9H20F3NO3S | 1 ML
SDP

Catalog No. 5000282632
Encompass_Preferred

Tetraethylammonium trifluoromethanesulfonate is a quaternary ammonium salt supplied as a 10 mM solution (one mL) intended for research use. It serves as a convenient source of the tetraethylammonium cation for small-scale experiments, phase-transfer catalysis, and method development in organic synthesis and analytical workflows.

  • Supplied as a 10 mM solution in one mL for ready-to-use small-volume experiments.
  • High purity (98.0%) suitable for research applications.
  • Provides tetraethylammonium cation for phase-transfer catalysis and ion pairing.
  • Molecular formula C9H20F3NO3S and molecular weight 279.32 g·mol⁻¹.
  • For research use only; not for human or diagnostic use.

Catalog No. 50-002-82632 Supplier Medchemexpress LLC Supplier No. HYW12767810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetraethylammonium trifluoromethanesulfonate is a quaternary ammonium salt supplied as a 10 mM solution (one mL) intended for research use. It serves as a convenient source of the tetraethylammonium cation for small-scale experiments, phase-transfer catalysis, and method development in organic synthesis and analytical workflows.

  • Supplied as a 10 mM solution in one mL for ready-to-use small-volume experiments.
  • High purity (98.0%) suitable for research applications.
  • Provides tetraethylammonium cation for phase-transfer catalysis and ion pairing.
  • Molecular formula C9H20F3NO3S and molecular weight 279.32 g·mol⁻¹.
  • For research use only; not for human or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.