Learn More
Medchemexpress LLC Tetraethylammonium trifluoromethanesulfonate | 35895-69-3 | MFCD00042586 | ≥98.0% | 279.32 g/mol | C9H20F3NO3S | 25 G
SDP
Catalog No. 5000282633
Encompass_Preferred
Tetraethylammonium trifluoromethanesulfonate is a quaternary ammonium salt used as a phase-transfer catalyst, supporting electrolyte, and reagent in organic synthesis and biochemical research. Supplied for research use only, it ishandled and stored sealed away from moisture per supplier guidance. CAS 35895-69-3; molecular weight 279.32 g/mol.
  • Quaternary ammonium salt useful as a phase-transfer catalyst.
  • Suitable as a supporting electrolyte in organic and electrochemical reactions.
  • High purity commonly reported by suppliers (≥98%).
  • Provided in research-scale packaging for laboratory use.
  • Store sealed and dry; recommended refrigerated storage for long-term stability.
Catalog No. 50-002-82633 Supplier Medchemexpress LLC Supplier No. HYW12767825G
Tetraethylammonium trifluoromethanesulfonate is a quaternary ammonium salt used as a phase-transfer catalyst, supporting electrolyte, and reagent in organic synthesis and biochemical research. Supplied for research use only, it ishandled and stored sealed away from moisture per supplier guidance. CAS 35895-69-3; molecular weight 279.32 g/mol.
  • Quaternary ammonium salt useful as a phase-transfer catalyst.
  • Suitable as a supporting electrolyte in organic and electrochemical reactions.
  • High purity commonly reported by suppliers (≥98%).
  • Provided in research-scale packaging for laboratory use.
  • Store sealed and dry; recommended refrigerated storage for long-term stability.