Learn More
Medchemexpress LLC Tetraethylene glycol | 10g
SDP
Catalog No. 5000202881
Encompass_Preferred
Tetraethylene glycol monotosylate is a cleavable and acylhydrazone-based ADC linker used in the synthesis of antibody-drug conjugates (ADCs)[1] Tetraethylene glycol monotosylate also can be used as a PROTAC linker that can be used in the synthesis of PROTACs
Catalog No. 50-002-02881 Supplier Medchemexpress LLC Supplier No. HY4154110G
Tetraethylene glycol monotosylate is a cleavable and acylhydrazone-based ADC linker used in the synthesis of antibody-drug conjugates (ADCs)[1] Tetraethylene glycol monotosylate also can be used as a PROTAC linker that can be used in the synthesis of PROTACs