Learn More
Medchemexpress LLC Tetraethylene glycol monomethyl ether | 23783-42-8 | MFCD00041756 | 99.6% | 208.25 | C9H20O5 | 25 G
SDP
Catalog No. 5000304168
Encompass_Preferred
Tetraethylene glycol monomethyl ether is a polyethylene glycol (PEG)-based linker and solvent used in medicinal chemistry and PROTAC synthesis. It is a clear, low-volatility liquid (C9H20O5, MW 208.25) supplied for research use; high purity supports consistent coupling and solubility enhancement in small-molecule syntheses.
  • Polyethylene glycol (PEG) spacer for linker design.
  • High purity (~99.6%) for reproducible reactions.
  • Improves solubility of hydrophobic fragments.
  • Compatible with common organic solvents and coupling chemistries.
  • Available in multiple pack sizes for scale flexibility.
Catalog No. 50-003-04168 Supplier Medchemexpress LLC Supplier No. 25GHYW018365
Tetraethylene glycol monomethyl ether is a polyethylene glycol (PEG)-based linker and solvent used in medicinal chemistry and PROTAC synthesis. It is a clear, low-volatility liquid (C9H20O5, MW 208.25) supplied for research use; high purity supports consistent coupling and solubility enhancement in small-molecule syntheses.
  • Polyethylene glycol (PEG) spacer for linker design.
  • High purity (~99.6%) for reproducible reactions.
  • Improves solubility of hydrophobic fragments.
  • Compatible with common organic solvents and coupling chemistries.
  • Available in multiple pack sizes for scale flexibility.