Learn More
Medchemexpress LLC Tetraethylene glycol monomethyl ether | 23783-42-8 | MFCD00041756 | 99.6% | 208.25 g/mol | C9H20O5 | 50 G
SDP
Catalog No. 5000301434
Encompass_Preferred
Tetraethylene glycol monomethyl ether is a polyethylene glycol (PEG)-based monomethyl ether used as a linker and building block in medicinal chemistry and PROTAC synthesis. It is a colorless to light yellow liquid with a moderate density and high reported purity, supplied in laboratory-scale pack sizes for synthetic applications.
  • PEG-based monomethyl ether useful as a PROTAC linker and synthetic building block.
  • High purity (99.61%) suitable for research-grade synthesis.
  • Molecular weight 208.25 g/mol, enabling predictable spacer length in conjugates.
  • Liquid, colorless to light yellow, easy to dispense and handle.
  • Available in multiple pack sizes, including 50 g, for bench-scale use.
  • Stable when stored under recommended temperature conditions.
Catalog No. 50-003-01434 Supplier Medchemexpress LLC Supplier No. HYW01836550G
Tetraethylene glycol monomethyl ether is a polyethylene glycol (PEG)-based monomethyl ether used as a linker and building block in medicinal chemistry and PROTAC synthesis. It is a colorless to light yellow liquid with a moderate density and high reported purity, supplied in laboratory-scale pack sizes for synthetic applications.
  • PEG-based monomethyl ether useful as a PROTAC linker and synthetic building block.
  • High purity (99.61%) suitable for research-grade synthesis.
  • Molecular weight 208.25 g/mol, enabling predictable spacer length in conjugates.
  • Liquid, colorless to light yellow, easy to dispense and handle.
  • Available in multiple pack sizes, including 50 g, for bench-scale use.
  • Stable when stored under recommended temperature conditions.