Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetraethylene glycol monotosylate | 77544-60-6 | MFCD22056303 | >97.0% | 348.41 g/mol | C15H24O7S | 1 G
SDP

Catalog No. 5000202761
Encompass_Preferred

Tetraethylene glycol monotosylate is a tosylated tetraethylene glycol reagent used as a cleavable linker or spacer in bioconjugation and organic synthesis. It is commonly used for antibody-drug conjugate (ADC) linker construction and related medicinal chemistry applications.

  • tosylated tetraethylene glycol structure provides a reactive leaving group for conjugation
  • cleavable acylhydrazone-compatible spacer for ADC and PROTAC synthesis
  • molecular weight 348.41 g/mol and formula C15H24O7S
  • CAS number 77544-60-6 for unambiguous identification
  • high purity (≥97%) suitable for synthetic applications
  • storage in pure form at -20°C or 4°C extends shelf life

Catalog No. 50-002-02761 Supplier Medchemexpress LLC Supplier No. HY415411G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetraethylene glycol monotosylate is a tosylated tetraethylene glycol reagent used as a cleavable linker or spacer in bioconjugation and organic synthesis. It is commonly used for antibody-drug conjugate (ADC) linker construction and related medicinal chemistry applications.

  • tosylated tetraethylene glycol structure provides a reactive leaving group for conjugation
  • cleavable acylhydrazone-compatible spacer for ADC and PROTAC synthesis
  • molecular weight 348.41 g/mol and formula C15H24O7S
  • CAS number 77544-60-6 for unambiguous identification
  • high purity (≥97%) suitable for synthetic applications
  • storage in pure form at -20°C or 4°C extends shelf life

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.