Learn More
Medchemexpress LLC Tetraethylenepentamine (pentahydrochloride) | 4961-41-5 | 99.8% | 250 MG
SDP
Catalog No. 5000413941
Encompass_Preferred
Tetraethylenepentamine pentahydrochloride is an effective chelator that increases the urinary excretion of copper. It is intended for research use only.
  • Effective chelator in increasing the urinary excretion of copper.
Catalog No. 50-004-13941 Supplier Medchemexpress LLC Supplier No. 250MGHYB2153
Tetraethylenepentamine pentahydrochloride is an effective chelator that increases the urinary excretion of copper. It is intended for research use only.
  • Effective chelator in increasing the urinary excretion of copper.