Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylgermanium, 99%

Catalog No. AAH2762506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2762506 5 g
AAH2762503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2762506 Supplier Thermo Scientific Chemicals Supplier No. H2762506
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 597-63-7 | C8H20Ge | 188.878 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 597-63-7
Molecular Formula C8H20Ge
Molecular Weight (g/mol) 188.878
MDL Number MFCD00015096
InChI Key QQXSEZVCKAEYQJ-UHFFFAOYSA-N
Synonym tetraethylgermanium, germane, tetraethyl, tetraethyl germane, germanium, tetraethyl, acmc-209mfg, c2h5 4ge, qqxsezvckaeyqj-uhfffaoysa, wln: 2-ge-2&2&2
PubChem CID 11703
IUPAC Name tetraethylgermane
SMILES CC[Ge](CC)(CC)CC

Specifications

Density 0.998
Boiling Point 163°C to 164°C
Flash Point 35°C (95°F)
Refractive Index 1.442
Quantity 5 g
UN Number UN1993
Formula Weight 188.88
Percent Purity 99%
Chemical Name or Material Tetraethylgermanium
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 209-905-7
RTECSNumber LY5290000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.