Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylphosphonium Bromide 98.0+%, TCI America™
SDP

Catalog No. T13795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13795G 5 g
T137925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13795G Supplier TCI America Supplier No. T13795G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4317-07-1
Molecular Formula C8H20BrP
Molecular Weight (g/mol) 227.13
MDL Number MFCD00054230
InChI Key LIXPXSXEKKHIRR-UHFFFAOYSA-M
PubChem CID 9859378
IUPAC Name tetraethylphosphanium;bromide
SMILES CC[P+](CC)(CC)CC.[Br-]

Specifications

Color White
Quantity 5 g
Formula Weight 227.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraethylphosphonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.