Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylphosphonium Hexafluorophosphate 99.0+%, TCI America™
SDP

Catalog No. T17465G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T17465G Supplier TCI America Supplier No. T17465G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 111928-07-5
Molecular Formula C8H20F6P2
Molecular Weight (g/mol) 292.19
MDL Number MFCD01631313
InChI Key WJZBUQKNTCKXAE-UHFFFAOYSA-N
PubChem CID 2760564
IUPAC Name hexafluoro-λ⁵-phosphanuide; tetraethylphosphanium
SMILES F[P-](F)(F)(F)(F)F.CC[P+](CC)(CC)CC

Specifications

Color White
Quantity 5 g
Formula Weight 292.19
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraethylphosphonium Hexafluorophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.