Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylsilane, 97%

Catalog No. AAL0332822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0332822 100 g
AAL0332814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0332822 Supplier Thermo Scientific Chemicals Supplier No. L0332822
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 631-36-7 | C8H20Si | 144.333 g/mol

Used as the intermediates of OLED, pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as the intermediates of OLED, pharmaceuticals.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-36-7
Molecular Formula C8H20Si
Molecular Weight (g/mol) 144.333
MDL Number MFCD00009019
InChI Key VCZQFJFZMMALHB-UHFFFAOYSA-N
Synonym silane, tetraethyl, tetraethylsilicon, tetraethylsilane, acmc-209ndb, c2h5 4si
PubChem CID 12426
IUPAC Name tetraethylsilane
SMILES CC[Si](CC)(CC)CC

Specifications

Boiling Point 153°C to 155°C
Melting Point -82°C
Quantity 100 g
Flash Point 26°C (78°F)
Odor Recognizable
Refractive Index 1.426
Density 0.767
UN Number UN1993
Beilstein 1732768
Solubility Information Insoluble in water.
Formula Weight 144.33
Percent Purity 97%
Chemical Name or Material Tetraethylsilane
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 211-155-0
RTECSNumber VV5500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.