Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrafluoro-1,4-benzoquinone 98.0+%, TCI America™
SDP

Catalog No. T07905G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07905G 5 g
T07901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T07905G Supplier TCI America Supplier No. T07905G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 527-21-9
Molecular Formula C6F4O2
Molecular Weight (g/mol) 180.06
MDL Number MFCD00001592
InChI Key JKLYZOGJWVAIQS-UHFFFAOYSA-N
Synonym tetrafluoro-1,4-benzoquinone, fluoranil, p-fluoranil, tetrafluoroquinone, tetrafluoro-p-benzoquinone, fluoroanil, tetrafluorocyclohexa-2,5-diene-1,4-dione, 2,5-cyclohexadiene-1,4-dione, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluoro-1,4-benzoquinone, 2,3,5,6-tetrafluoro 1,4 benzoquinone
PubChem CID 68239
IUPAC Name tetrafluorocyclohexa-2,5-diene-1,4-dione
SMILES FC1=C(F)C(=O)C(F)=C(F)C1=O

Specifications

Quantity 5 g
Formula Weight 180.06
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrafluoro-1,4-benzoquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.