Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrafluoro-2-(tetrafluoro-2-iodoethoxy)ethanesulfonyl Fluoride (stabilized with Na2S2O3) 95.0+%, TCI America™
SDP

Catalog No. T29145G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T29145G Supplier TCI America Supplier No. T29145G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 66137-74-4
Molecular Formula C4F9IO3S
Molecular Weight (g/mol) 425.991
MDL Number MFCD00798139
InChI Key XSLYISNQTJHKMP-UHFFFAOYSA-N
PubChem CID 2775169
IUPAC Name 1,1,2,2-tetrafluoro-2-(1,1,2,2-tetrafluoro-2-iodoethoxy)ethanesulfonyl fluoride
SMILES C(C(F)(F)S(=O)(=O)F)(OC(C(F)(F)I)(F)F)(F)F

Specifications

Color Red
Boiling Point 127°C
Quantity 5 g
UN Number 3265
Formula Weight 425.99
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetrafluoro-2-(tetrafluoro-2-iodoethoxy)ethanesulfonyl Fluoride (stabilized with Na2S2O3)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.