Learn More
AdipoGen Tetrafluoro-IBA
SDP
Catalog No. 502560662
Encompass_Preferred
Chemical. CAS 954373-94-5. Formula C7HF4IO3. MW 335.98. Synthetic Hypervalent iodine reagents. Convenient substitute of o-Iodosobenzoic acid IBA as an oxidant. Can be used for installations of alpha/beta-unsaturation to carbonyl compounds, oxidations and cyclizations by single-electron transfer processes. Promotes faster reactions than IBA with better solubility in common organic solvents. An attractive feature is the ability to directly measure reactivity by 19F NMR spectroscopy without the use of deuterated solvents.
Catalog No. 50-256-0662 Supplier AdipoGen Supplier No. CDXI0027G005
Chemical. CAS 954373-94-5. Formula C7HF4IO3. MW 335.98. Synthetic Hypervalent iodine reagents. Convenient substitute of o-Iodosobenzoic acid IBA as an oxidant. Can be used for installations of alpha/beta-unsaturation to carbonyl compounds, oxidations and cyclizations by single-electron transfer processes. Promotes faster reactions than IBA with better solubility in common organic solvents. An attractive feature is the ability to directly measure reactivity by 19F NMR spectroscopy without the use of deuterated solvents.