Learn More
AdipoGen Tetrafluoro-IBX
SDP
Catalog No. 502560660
Encompass_Preferred
Chemical. CAS 954373-95-6. Formula C7HF4IO4. MW 352. Synthetic. Convenient substitute of o-iodoxybenzoic acid IBX as an oxidant. FIBX is safer, has neutral properties behaves as an equivalent to IBX in the oxidation of alcohols to ketones or aldehydes in polar solvents and shows better solubility.
Catalog No. 50-256-0660 Supplier AdipoGen Supplier No. CDXI0028M500
Chemical. CAS 954373-95-6. Formula C7HF4IO4. MW 352. Synthetic. Convenient substitute of o-iodoxybenzoic acid IBX as an oxidant. FIBX is safer, has neutral properties behaves as an equivalent to IBX in the oxidation of alcohols to ketones or aldehydes in polar solvents and shows better solubility.