Learn More
Strem, An Ascensus Company CAS# 16872-11-0. 500g. Tetrafluoroboric acid, 48% aqueous solution. MFCD00011345
SDP
Catalog No. 5090113705
Encompass
CAS# 16872-11-0. 500g. Tetrafluoroboric acid, 48% aqueous solution. MFCD00011345. Molecular Weight: 87.81. Molecular Formula: HBF4. Color/form: colorless to light yellow liq. Strem# 93-0503. http://www.strem.com/catalog/v/93-0503/
Catalog No. 50-901-13705 Supplier Strem, An Ascensus Company Supplier No. 930503500G
CAS# 16872-11-0. 500g. Tetrafluoroboric acid, 48% aqueous solution. MFCD00011345. Molecular Weight: 87.81. Molecular Formula: HBF4. Color/form: colorless to light yellow liq. Strem# 93-0503. http://www.strem.com/catalog/v/93-0503/