Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrafluoroboric Acid Diethyl Ether Complex

Catalog No. 400068400ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
400 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-006-8400ML 400 mL
40-006-8100ML 100 mL
40-006-825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 40-006-8400ML Supplier Sigma Aldrich Supplier No. 400068400ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetrafluoroboric acid diethyl ether complex can be used to facilitate phosphine-borane decomplexation from bisphosphine-borane complexes for the preparation of bisphosphine ligands. It can also act as ligands to synthesize with transition-metal complexes.

Specifications

CAS 67969-82-8
Density 1.19 g/mL at 20°C, 1.18 g/mL at 25°C
Molecular Formula C4H11BF4O
Linear Formula HBF4 -+ O(CH2CH3)2
MDL Number MFCD00085352
Quantity 400 mL
Synonym Fluoroboric acid diethyl ether complex
Molecular Weight (g/mol) 161.93 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.