Tetrafluoroboric Acid, Liquid, 48% min., Alfa Aesar™

Manufacturer: Alfa Aesar 1148436

Quantity 500g

 View more versions of this product

Catalog No. AA1148436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16872-11-0
    Physical Form Liquid
    Assay Percent Range 48% min.
    Molecular Formula HBF4
    Formula Weight 87.81
    MDL Number MFCD00011345
    Chemical Name or Material Tetrafluoroboric Acid

    Specifications

    Formula Weight 87.81
    Quantity 500g
    DOT Information DOT Class: 8, Packaging Group: II
    EINECS Number 240-898-3
    Physical Form Liquid
    Assay Percent Range 48% min.
    Molecular Formula HBF4
    MDL Number MFCD00011345
    CAS 16872-11-0
    Health Hazard 1 H314-H227: Causes severe skin burns and eye damage. Combustible liquid.
    Chemical Name or Material Tetrafluoroboric Acid

    H314-H227: Causes severe skin burns and eye damage. Combustible liquid.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit