Learn More
Sigma Aldrich Fine Chemicals Biosciences Tetrafluoroboric acid solu500G
Catalog No. 502897843
Encompass
Tetrafluoroboric acid is used as a catalyst for the protection and deprotection reactions of various carbohydrates. It participates in the synthesis of 4-sulfonic acid phenyl diazonium tetrafluoroborate which was required for the preparation of sulfonated graphene (SG).
Catalog No. 50-289-7843 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 207934500G
Tetrafluoroboric acid is used as a catalyst for the protection and deprotection reactions of various carbohydrates. It participates in the synthesis of 4-sulfonic acid phenyl diazonium tetrafluoroborate which was required for the preparation of sulfonated graphene (SG).