Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrafluoroisophthalamide 94.0+%, TCI America™
SDP

Catalog No. T18511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18511G 1 g
T18515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18511G Supplier TCI America Supplier No. T18511G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 153279-27-7
Molecular Formula C8H4F4N2O2
Molecular Weight (g/mol) 236.126
InChI Key JDNPZWSPPAVPPP-UHFFFAOYSA-N
Synonym Tetrafluoro-m-phthalamide
PubChem CID 11172398
IUPAC Name 2,4,5,6-tetrafluorobenzene-1,3-dicarboxamide
SMILES C1(=C(C(=C(C(=C1F)F)F)C(=O)N)F)C(=O)N

Specifications

Color White
Quantity 1 g
Formula Weight 236.13
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrafluoroisophthalamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.