Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrafluorophthalonitrile 98.0+%, TCI America™
SDP

Catalog No. T14025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14025G 5 g
T140225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14025G Supplier TCI America Supplier No. T14025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1835-65-0
Molecular Formula C8F4N2
Molecular Weight (g/mol) 200.096
MDL Number MFCD00001774
InChI Key OFLRJMBSWDXSPG-UHFFFAOYSA-N
Synonym tetrafluorophthalonitrile, 3,4,5,6-tetrafluorophthalonitrile, 1,2-dicyano-3,4,5,6-tetrafluorobenzene, 1,2-benzenedicarbonitrile, 3,4,5,6-tetrafluoro, 1,2-benzenedicarbonitrile, 3,4,5,6-tetrafluoro-;3,4,5,6-tetrafluorobenzene-1,2-dicarbonitrile, pubchem4564, acmc-209ekr, ksc496c0b, 3,4,5,6-tetrafluorophthalonitrile #
PubChem CID 74600
IUPAC Name 3,4,5,6-tetrafluorobenzene-1,2-dicarbonitrile
SMILES C(#N)C1=C(C(=C(C(=C1F)F)F)F)C#N

Specifications

Melting Point 87°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 200.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrafluorophthalonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.