Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrafluoroterephthalonitrile, 98%

Catalog No. AAH6132614
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6132614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6132614 Supplier Thermo Scientific Chemicals Supplier No. H6132614
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1835-49-0 | C8F4N2 | 200.096 g/mol

Tetrafluoroterephthalonitrile can be used in the synthesis of Polymers of Intrinsic Micro porosity (PIM). It is used to study ultraviolet (UV)-rearranged polymers of PIM-1 membranes for efficient separation of H2 and CO2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrafluoroterephthalonitrile can be used in the synthesis of Polymers of Intrinsic Micro porosity (PIM). It is used to study ultraviolet (UV)-rearranged polymers of PIM-1 membranes for efficient separation of H2 and CO2.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1835-49-0
Molecular Formula C8F4N2
Molecular Weight (g/mol) 200.096
MDL Number MFCD00001776
InChI Key PCRSJGWFEMHHEW-UHFFFAOYSA-N
Synonym tetrafluoroterephthalonitrile, 2,3,5,6-tetrafluoroterephthalonitrile, diamond 2031, 2,3,5,6-tetrafluoro-1,4-benzenedicarbonitrile, 1,4-benzenedicarbonitrile, 2,3,5,6-tetrafluoro, 1,4-dicyanotetrafluorobenzene, terephthalonitrile, tetrafluoro, perfluoroterephthalonitrile, tetrafluoroterephthalodinitrile, tetrafluorobenzene-1,4-dicarbonitrile
PubChem CID 15783
IUPAC Name 2,3,5,6-tetrafluorobenzene-1,4-dicarbonitrile
SMILES C(#N)C1=C(C(=C(C(=C1F)F)C#N)F)F

Specifications

Melting Point 197°C to 199°C
Quantity 25 g
UN Number UN3439
Solubility Information Soluble in hot methanol.
Formula Weight 200.09
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Tetrafluoroterephthalonitrile
Hazard Category H301-H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 217-397-3
RTECSNumber CZ1955000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.