Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetraglycine | 637-84-3 | MFCD00008129 | 98.1% | 246.22 | C8H14N4O5 | 25 G
SDP

Catalog No. 5000282299
Encompass_Preferred

Tetraglycine is an oligopeptide composed of four glycine monomers, used in biochemical research to enrich glycine pools in plasma and tissues. It is supplied as a high-purity powder for laboratory research with storage recommendations to preserve stability and activity.

  • Oligopeptide of four glycine residues.
  • Molecular weight 246.22 g/mol.
  • CAS number 637-84-3.
  • Purity 98.1%.
  • Available in powder packages up to 25 g.
  • Recommended storage: powder sealed, away from moisture and light; -80°C for long term, -20°C for up to one year.
  • For research use only.

Catalog No. 50-002-82299 Supplier Medchemexpress LLC Supplier No. HYW14246725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetraglycine is an oligopeptide composed of four glycine monomers, used in biochemical research to enrich glycine pools in plasma and tissues. It is supplied as a high-purity powder for laboratory research with storage recommendations to preserve stability and activity.

  • Oligopeptide of four glycine residues.
  • Molecular weight 246.22 g/mol.
  • CAS number 637-84-3.
  • Purity 98.1%.
  • Available in powder packages up to 25 g.
  • Recommended storage: powder sealed, away from moisture and light; -80°C for long term, -20°C for up to one year.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.