Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetraheptylammonium iodide | 3535-83-9 | MFCD00041984 | >=99.0% | 537.69 | C28H60IN | 25 G
SDP

Catalog No. 5000282813
Encompass_Preferred

Tetraheptylammonium iodide is a quaternary ammonium salt supplied as a solid reagent for laboratory research use. It is commonly used in synthesis and biochemical assays and should be handled according to standard laboratory safety practices.

  • Solid form, white to light yellow.
  • Molecular formula C28H60IN.
  • Molecular weight 537.69.
  • High assay purity (reported ≥99.0% by some suppliers).
  • Packaged as 25 G for laboratory use.
  • Storage: sealed at 4°C, protect from moisture and light.

Catalog No. 50-002-82813 Supplier Medchemexpress LLC Supplier No. HYW12766125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetraheptylammonium iodide is a quaternary ammonium salt supplied as a solid reagent for laboratory research use. It is commonly used in synthesis and biochemical assays and should be handled according to standard laboratory safety practices.

  • Solid form, white to light yellow.
  • Molecular formula C28H60IN.
  • Molecular weight 537.69.
  • High assay purity (reported ≥99.0% by some suppliers).
  • Packaged as 25 G for laboratory use.
  • Storage: sealed at 4°C, protect from moisture and light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.