Learn More
AdipoGen Tetrahexylammonium HSO4
SDP
Supplier:  AdipoGen CDXT0610G025
Encompass_Preferred
Chemical. CAS 32503-34-7. Formula C24H53NO4S. MW 451.75. Tetrahexylammonium hydrogensulfate THAHS is a quaternary ammonium salt used in various chemical and biochemical applications. It consists of a tetracationic ammonium ion C6H134N and a hydrogen sulfate anion HSO4-. THAHS is used as a phase-transfer catalyst PCT and can facilitate various chemical reactions by aiding in the transfer of reactants between immiscible phases. Phase-transfer catalysis involves the transfer of reactants from one phase usually an aqueous phase to another phase usually an organic phase to facilitate a chemical reaction. THAHS can be employed in organic synthesis, particularly in reactions that involve the transfer of anions or reagents between organic and aqueous phases. Some common applications include, quaternization reactions, nucleophilic substitution reactions or halogenation reactions.
Catalog No. 50-255-9929