Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrahexylammonium Hydroxide Solution

Catalog No. 87306100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
87-306-100ML 100 mL
87-306-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 87-306-100ML Supplier Sigma Aldrich Supplier No. 87306100ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetrahexylammonium hydroxide is a phase-transfer reagent that can be used:To synthesize +a-D-xyloside- and +a-D-xylobioside-based ionic liquids containing tetrahexylammonium cation.As an alkali-free gelling agent to prepare amorphous microporous silica-alumina.To synthesize tetrahexylammonium prolinate, a promising candidate to prepare a solid supported ionic liquid phase (SILP) carbon dioxide absorber.

Specifications

Boiling Point 232°C
Molecular Formula C24H53NO
Linear Formula [CH3(CH2)5]4N(OH)
Quantity 100 mL
MDL Number MFCD00059255
Molecular Weight (g/mol) 371.68 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.