Learn More
Medchemexpress LLC Tetrahexylammonium iodide | 2138-24-1 | MFCD00041981 | ≥99.0% | 481.58 g/mol | C24H52IN | 25 G
SDP
Catalog No. 5000282743
Encompass_Preferred
Tetrahexylammonium iodide is a quaternary ammonium salt used as a reagent in organic synthesis and biochemical assays. It is commonly employed where tetraalkylammonium salts are required for phase-transfer catalysis, ion pairing, or as electrolyte components. Consult the product data sheet and SDS for handling, storage, and safety information.
Catalog No. 50-002-82743 Supplier Medchemexpress LLC Supplier No. HYW12766025G
Tetrahexylammonium iodide is a quaternary ammonium salt used as a reagent in organic synthesis and biochemical assays. It is commonly employed where tetraalkylammonium salts are required for phase-transfer catalysis, ion pairing, or as electrolyte components. Consult the product data sheet and SDS for handling, storage, and safety information.