Learn More
Medchemexpress LLC Tetrahydro-11-deoxycortisol | 68-60-0 | 98.2% | 350.49 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11342310MG
Encompass_Preferred
Tetrahydro-11-deoxycortisol (Tetrahydrodeoxycortisol) is an endogenous metabolite present in urine that can be used for the research of 11 Beta Hydroxylase Deficiency.
  • Endogenous metabolite
  • Used for the research of 11 Beta Hydroxylase Deficiency
  • Structure classification: Steroids
  • Initial source: Endogenous metabolite, disease markers: Cancer, infection
Catalog No. 50-004-37508