Learn More
Medchemexpress LLC Tetrahydro-2H-pyran-2-one | 542-28-9 | 99.9% | 100.12 | 10 MM * 1 ML
SDP
Catalog No. 5000451456
Encompass_Preferred
Tetrahydro-2H-pyran-2-one (δ-valerolactone) is an endogenous metabolite possessing antioxidant capacity. It functions as a small active molecule and can be utilized as a drug intermediate.
  • Endogenous metabolite with antioxidant capacity.
  • Small active molecule.
  • Can be used as a drug intermediate.
  • Generates polymers through ring-opening polymerization.
  • Polymerizes spontaneously to produce linear polyesters.
  • Can copolymerize with other monomers (such as ε-caprolactone) to produce copolymers.
Catalog No. 50-004-51456 Supplier Medchemexpress LLC Supplier No. HYW01299710MM/1ML
Tetrahydro-2H-pyran-2-one (δ-valerolactone) is an endogenous metabolite possessing antioxidant capacity. It functions as a small active molecule and can be utilized as a drug intermediate.
  • Endogenous metabolite with antioxidant capacity.
  • Small active molecule.
  • Can be used as a drug intermediate.
  • Generates polymers through ring-opening polymerization.
  • Polymerizes spontaneously to produce linear polyesters.
  • Can copolymerize with other monomers (such as ε-caprolactone) to produce copolymers.