Learn More
Medchemexpress LLC Tetrahydro-2H-pyran-2-one-d4 | 42932-61-6 | HY-W012997S | 98.6% | 104.14 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW012997S10MG
Encompass_Preferred
Tetrahydro-2H-pyran-2-one-d4 is the deuterium labeled Tetrahydro-2H-pyran-2-one. Tetrahydro-2H-pyran-2-one is an endogenous metabolite with antioxidant capacity and a small active molecule that can be used as a drug intermediate.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-51390