Learn More
Medchemexpress LLC Tetrahydro-2h-pyran-2-one (δ-valerolactone) | 542-28-9 | 99.94% | 100.12 | 25 G
SDP
Catalog No. 5000451382
Encompass_Preferred
Tetrahydro-2H-pyran-2-one (δ-valerolactone) is an endogenous metabolite with antioxidant capacity. It is a small active molecule that can be used as a drug intermediate. This compound generates polymers through ring-opening polymerization.
  • Can be used as a drug intermediate
  • Generates polymers through ring-opening polymerization
  • Polymerizes spontaneously to produce linear polyesters
  • Copolymerizes with other monomers to produce copolymers
Catalog No. 50-004-51382 Supplier Medchemexpress LLC Supplier No. 25GHYW012997
Tetrahydro-2H-pyran-2-one (δ-valerolactone) is an endogenous metabolite with antioxidant capacity. It is a small active molecule that can be used as a drug intermediate. This compound generates polymers through ring-opening polymerization.
  • Can be used as a drug intermediate
  • Generates polymers through ring-opening polymerization
  • Polymerizes spontaneously to produce linear polyesters
  • Copolymerizes with other monomers to produce copolymers