Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydro-2H-pyran-4-carbonyl chloride, ≥97%, Thermo Scientific™

Catalog No. CC29902CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC29902CB 250 mg
CC29902DA 1 g
CC29902DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC29902CB Supplier Thermo Scientific Chemicals Supplier No. CC29902CB
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 40191-32-0
Molecular Formula C6H9ClO2
Molecular Weight (g/mol) 148.59
MDL Number MFCD06200863
InChI Key RYGUCYSSMOFTSH-UHFFFAOYSA-N
Synonym tetrahydro-2h-pyran-4-carbonyl chloride, 2h-pyran-4-carbonyl chloride, tetrahydro, tetrahydro-2h-pyran-4-carbonylchloride, pubchem12437, ksc235m5r, tetrahydropyran-4-carbonyl chloride, tetrahydro-pyran-4-carbonyl chloride, 4-chloroformyl tetrahydro-2h-pyran, tetrahydropyran-4-carboxylic chloride, 4-chlorocarbonyl tetrahydro-2h-pyran
PubChem CID 2795505
IUPAC Name oxane-4-carbonyl chloride
SMILES ClC(=O)C1CCOCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 148.59
Percent Purity 95%
Chemical Name or Material Tetrahydro-2H-pyran-4-carbonyl chloride
Hazard Category Danger
Hazard Statement GHS P Statement Causes severe skin burns and eye damage. Reacts violently with water. Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair):
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.