Learn More
Medchemexpress LLC Tetrahydro-2H-pyran-4-ol-d5 | 1219795-46-6 | 107.16 | 1 MG
SDP
Catalog No. 5000447958
Encompass_Preferred
Tetrahydro-2H-pyran-4-ol-d5 is a deuterium labeled compound designed for research applications. It serves as a tracer and an internal standard for quantitative analysis methods such as NMR, GC-MS, and LC-MS. Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Store in pure form at -20°C for 3 years.
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month.
  • Ships at room temperature in continental US (may vary elsewhere).
Catalog No. 50-004-47958 Supplier Medchemexpress LLC Supplier No. HYW004264S1MG
Tetrahydro-2H-pyran-4-ol-d5 is a deuterium labeled compound designed for research applications. It serves as a tracer and an internal standard for quantitative analysis methods such as NMR, GC-MS, and LC-MS. Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Store in pure form at -20°C for 3 years.
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month.
  • Ships at room temperature in continental US (may vary elsewhere).