Learn More
Medchemexpress LLC Tetrahydro-2h-pyran-4-ol-d5 | 1219795-46-6 | 107.16 | 10 MG
SDP
Catalog No. 5000445711
Encompass_Preferred
Tetrahydro-2H-pyran-4-ol-d5 is a deuterium labeled compound intended for research use only. It serves as a tracer and an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Deuterium labeling can influence pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-45711 Supplier Medchemexpress LLC Supplier No. HYW004264S10MG
Tetrahydro-2H-pyran-4-ol-d5 is a deuterium labeled compound intended for research use only. It serves as a tracer and an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Deuterium labeling can influence pharmacokinetic and metabolic profiles of drugs