Learn More
Accela Chembio Inc MF: C5H10O2 | MW: 102.13 | >97% | BP: 183? at 760 mmHg | Density: 1.071.
Catalog No. 50249905
Encompass_Preferred
MF: C5H10O2 | MW: 102.13 | >97% | BP: 183? at 760 mmHg | Density: 1.071.
Catalog No. 50-249-905 Supplier Accela Chembio Inc Supplier No. SY00268725G
MF: C5H10O2 | MW: 102.13 | >97% | BP: 183? at 760 mmHg | Density: 1.071.