Learn More
Medchemexpress LLC Tetrahydro-4H-pyran-4-one | 29943-42-8 | 100.0% | C5H8O2 | 10 MM 1 ML
SDP
Catalog No. 5000434101
Encompass_Preferred
Tetrahydro-4H-pyran-4-one is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is employed in the preparation of 4-methoxytetrahydropyran-4-yl protecting group, the synthesis of symmetric tetra substituted methanes, and is a useful protecting agent for alcohols in nucleotide synthesis.
  • Used in preparation of 4-methoxytetrahydropyran-4-yl protecting group.
  • Useful protecting agent for alcohols in nucleotide synthesis.
  • Employed in studies of enantioselective alpha-aminoxylation of ketones.
  • Undergoes condensation reactions in the preparation of dipeptides and spiroimidazolones.
  • Utilized in Wittig reactions for the synthesis of penicillins and in a ring of vitamin D3.
Catalog No. 50-004-34101 Supplier Medchemexpress LLC Supplier No. HY7648710MM/1ML
Tetrahydro-4H-pyran-4-one is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is employed in the preparation of 4-methoxytetrahydropyran-4-yl protecting group, the synthesis of symmetric tetra substituted methanes, and is a useful protecting agent for alcohols in nucleotide synthesis.
  • Used in preparation of 4-methoxytetrahydropyran-4-yl protecting group.
  • Useful protecting agent for alcohols in nucleotide synthesis.
  • Employed in studies of enantioselective alpha-aminoxylation of ketones.
  • Undergoes condensation reactions in the preparation of dipeptides and spiroimidazolones.
  • Utilized in Wittig reactions for the synthesis of penicillins and in a ring of vitamin D3.