Learn More
Medchemexpress LLC Tetrahydro-4H-pyran-4-one | 29943-42-8 | >99.9% | C5H8O2 | 1 KG
SDP
Catalog No. 5000434640
Encompass_Preferred
Tetrahydro-4H-pyran-4-one is a biochemical reagent used as a biological material or organic compound for life science related research. It is employed in the preparation of 4-methoxytetrahydropyran-4-yl protecting groups and the synthesis of symmetric tetra substituted methanes. The methyl enol ether is a useful protecting agent for alcohols, particularly in nucleotide synthesis, offering advantages over 3,4-Dihydro-2H-pyran. It also undergoes condensation reactions and is utilized in Wittig reactions.
  • Used in the preparation of protecting groups.
  • Key in the synthesis of symmetric tetra substituted methanes.
  • Acts as a protecting agent for alcohols.
  • Involved in condensation reactions for dipeptides and spiroimidazolones.
  • Utilized in Wittig reactions for penicillin and vitamin D3 ring synthesis.
Catalog No. 50-004-34640 Supplier Medchemexpress LLC Supplier No. HY764871KG
Add to Cart
Add to Cart
Tetrahydro-4H-pyran-4-one is a biochemical reagent used as a biological material or organic compound for life science related research. It is employed in the preparation of 4-methoxytetrahydropyran-4-yl protecting groups and the synthesis of symmetric tetra substituted methanes. The methyl enol ether is a useful protecting agent for alcohols, particularly in nucleotide synthesis, offering advantages over 3,4-Dihydro-2H-pyran. It also undergoes condensation reactions and is utilized in Wittig reactions.
  • Used in the preparation of protecting groups.
  • Key in the synthesis of symmetric tetra substituted methanes.
  • Acts as a protecting agent for alcohols.
  • Involved in condensation reactions for dipeptides and spiroimidazolones.
  • Utilized in Wittig reactions for penicillin and vitamin D3 ring synthesis.